missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Fodrin Mouse anti-Human, Clone: SPTAN1/3352, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07267-100ug
This item is not returnable.
View return policy
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Alpha Fodrin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| Recombinant fragment of human Alpha Fodrin protein (around aa 2351-2475) (exact sequence is proprietary)) | |
| 100 μg | |
| Neuroscience | |
| 6709 | |
| Store at 4°C. | |
| IgG2c κ |
| ELISA, Peptide Array | |
| SPTAN1/3352 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction