missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Gal Epitope Antibody (m86) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20112
This item is not returnable.
View return policy
Description
alpha-Gal Epitope Monoclonal antibody specifically detects alpha-Gal Epitope in Epitope Tag samples. It is validated for Flow Cytometry, ELISA
Specifications
| alpha-Gal Epitope | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Tag | |
| Purified |
| Flow Cytometry, ELISA | |
| m86 | |
| Unconjugated | |
| 3-alpha-galactobiose, 3-O-alpha-D-Galactopyranosyl-D-galactopyranose, 3-O-alpha-D-galactosylgalactose, alpha 1-3 galactobiose, Bdi-epitope, Gal(alpha)1-3Gal(beta)1-4GlcNAc-R, Gal-1-3-Gal, Galactose-alpha-1,3-galactose, Gal-alpha-1-3-Gal, Galili antigen | |
| Rabbit red cells, on which alpha-gal is the major carbohydrate epitope, were injected into alpha 1,3 galactosyltransferase knock-out mice. | |
| 0.05 mg | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction