missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Galactosidase A/GLA Antibody (30H11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26552-100ul
This item is not returnable.
View return policy
Description
alpha-Galactosidase A/GLA Monoclonal antibody specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| alpha-Galactosidase A/GLA | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 30H11 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Agalsidase, agalsidase alfa, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase | |
| A synthesized peptide derived from Human alpha-Galactosidase A/GLA [UniProt P06280] | |
| 100 μL | |
| Cardiovascular Biology | |
| 2717 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction