missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha-II Spectrin Polyclonal Antibody

Description
This antibody contains enough material to conduct 10 mini-Western blots.
SPTAN1 (spectrin alpha chain, non-erythrocytic 1) is a part of the spectrin family of filamentous cytoskeletal proteins. They function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. SPTAN1 has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | alpha-II Spectrin |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with no preservative |
| Gene | SPTAN1 |
| Gene Accession No. | P16086, P16546, Q13813 |
| Gene Alias | (ALPHA)II-SPECTRIN; 2610027H02Rik; A2a; a--fodrin; alpha; alpha II spectrin; alpha-fodrin; alpha-II spectrin; alpha-spectrin; alpha-spectrin 2; alpha-spectrin 2, brain; brain; brain alpha-spectrin; EIEE5; FLJ44613; Fodrin alpha chain; inhibitory protein factor; IPF; NEAS; noerythroid alpha-spectrin 2; Non erythrocytic spectrin alpha; non-erythrocytic 1 (alpha-fodrin); non-erythroid alpha chain; nonerythroid alpha-spectrin; nonerythroid alpha-spectrin 2; putative alpha-spectrin; SPECA; Spectrin; Spectrin alpha; spectrin alpha chain; Spectrin alpha chain brain; spectrin alpha chain, brain; spectrin alpha chain, non-erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; Spna2; Spna-2; Spta2; Sptan1 |
| Gene Symbols | SPTAN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?