Learn More
alpha-Internexin Antibody (S03-6H1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19567-50ul
 View more versions of this product
Product Code. 18672455
  • € 421.00 / 50µL
Description
alpha-Internexin Monoclonal antibody specifically detects alpha-Internexin in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Internexin
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S03-6H1
Unconjugated
66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1
A synthetic peptide of human alpha-Internexin (Uniprot # Q16352)
50 μg
Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
9118
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.