missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Internexin Monoclonal Antibody (1D2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA316752
This item is not returnable.
View return policy
Description
alpha Internexin Monoclonal Antibody for Western Blot, IHC (Free), ICC/IF
alpha-Internexin is a Class IV intermediate filament subunit expressed very early in neuronal development. It is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. Many classes of mature neurons contain a-internexin in addition to NF-L, NF-M and NF-H. In some mature neurons a-internexin is the only neurofilament subunit expressed. Antibodies to internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition the very early developmental expression of a-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cell committed to the neuronal lineage. The use of antibodies to this protein in the study of brain tumors has not been examined to date, but is likely to be of interest.
Specifications
| alpha Internexin | |
| Monoclonal | |
| 1 | |
| PBS with 50% glycerol and 0.035% sodium azide | |
| P23565, P46660, Q16352 | |
| INA | |
| Purified recombinant rat alpha-internexin expressed in and purified from E. coli. | |
| 500 μL | |
| Primary | |
| Feline, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Free Floating), Western Blot, Immunocytochemistry | |
| 1D2 | |
| Unconjugated | |
| INA | |
| 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 101085925, 226180, 24503, 9118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction