missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Internexin Polyclonal Antibody

Description
In Western blot, a prominent band at ~66 kDa is observed, as well as smaller lower bands which are apparently degradation products. A minor band at ~150 kDa is also seen, resulting from dimerization of alpha-internexin.
alpha-Internexin is a Class IV intermediate filament subunit expressed very early in neuronal development. It is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. Many classes of mature neurons contain a-internexin in addition to NF-L, NF-M and NF-H. In some mature neurons a-internexin is the only neurofilament subunit expressed. Antibodies to internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition the very early developmental expression of a-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cell committed to the neuronal lineage. The use of antibodies to this protein in the study of brain tumors has not been examined to date, but is likely to be of interest.
Specifications
Specifications
| Antigen | alpha Internexin |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 5mM sodium azide |
| Gene | INA |
| Gene Accession No. | P23565, P46660, Q08DH7, Q16352 |
| Gene Alias | 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 |
| Gene Symbols | INA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?