missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylacyl-CoA Racemase/AMACR Antibody (CL9362) [DyLight 650], Novus Biologicals™

Product Code. 30525737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525737 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525737 Supplier Novus Biologicals Supplier No. NBP343876C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Methylacyl-CoA Racemase/AMACR Monoclonal antibody specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Methylacyl-CoA Racemase/AMACR
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL9362
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q9UHK6, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Prostate Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23600
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.