Learn More
alpha-Methylstyrene-d10, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | alpha-Methylstyrene-d10 |
| CAS | 10362-82-0 |
| Molecular Formula | C9 D10 |
| Quantity | 0.1 g |
| Synonym | Styrene-β,β,2,3,4,5,6-d7, α-methyl-d3- (8 CI), α-(Methyl-d3)styrene-β,β,2,3,4,5,6-d7, [1-(Methyl-d3)ethenyl-2,2-d2]benzene-2,3,4,5,6-d5, α-Methylstyrene-d10, α-Methylstyrene-D10 |
| InChI Formula | InChI=1 S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7 H,1H2,2H3/i1D2,2D3,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]C(=C(c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H])C([2 H])([2 H])[2 H])[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(1,1,3,3,3-pentadeuterioprop-1-en-2-yl)benzene |
| Molecular Weight (g/mol) | 128.237 |
| Formula Weight | 128.141 g/mol |
| Show More |