missing translation for 'onlineSavingsMsg'
Learn More

alpha-N-terminal Methyltransferase 1A/METTL11A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-12704-100ul

 View more versions of this product

Product Code. 18601307

  • € 322.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

alpha-N-terminal Methyltransferase 1A/METTL11A Polyclonal antibody specifically detects alpha-N-terminal Methyltransferase 1A/METTL11A in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

alpha-N-terminal Methyltransferase 1A/METTL11A
Polyclonal
Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000
AD-003, alpha N-terminal protein methyltransferase 1A, C9orf32, chromosome 9 open reading frame 32, EC 2.1.1.-, methyltransferase like 11A, Methyltransferase-like protein 11A, NRMT, N-terminal RCC1 methyltransferase, NTM1A, NTMT1, X-Pro-Lys N-terminal protein methyltransferase 1A
Produced in rabbits immunized with E. coli-derived Human alpha-N-terminal Methyltransferase 1A/METTL11A fragment
100 μg
Cell Biology, Cell Cycle and Replication
28989
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.