missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-N-terminal Methyltransferase 1A/METTL11A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12704-50ul
This item is not returnable.
View return policy
Description
alpha-N-terminal Methyltransferase 1A/METTL11A Polyclonal antibody specifically detects alpha-N-terminal Methyltransferase 1A/METTL11A in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| alpha-N-terminal Methyltransferase 1A/METTL11A | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| AD-003, alpha N-terminal protein methyltransferase 1A, C9orf32, chromosome 9 open reading frame 32, EC 2.1.1.-, methyltransferase like 11A, Methyltransferase-like protein 11A, NRMT, N-terminal RCC1 methyltransferase, NTM1A, NTMT1, X-Pro-Lys N-terminal protein methyltransferase 1A | |
| Produced in rabbits immunized with E. coli-derived Human alpha-N-terminal Methyltransferase 1A/METTL11A fragment | |
| 50 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 28989 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction