Learn More
alpha-Smooth Muscle Actin Antibody (1A4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1420Y
Product Code. 30066626
  • ONLINE EXCLUSIVE PRICE
    € 674.00
    € 637.35 / 0.10mL
    Save € 36.65
    5% Off
Description
alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Smooth Muscle Actin
Monoclonal
DyLight 680
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
1A4
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC
AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Peptide corresponding to residues EEEDSTALVC of the N-term of human alpha-smooth muscle actin (1)
0.1 mL
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
59
Store at 4C in the dark.