missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Antibody (1A4) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1420JF635
This item is not returnable.
View return policy
Description
alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF,Dual RNAscope ISH-IHC
Specifications
| alpha-Smooth Muscle Actin | |
| Monoclonal | |
| Janelia Fluor 635 | |
| AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
| Peptide corresponding to residues EEEDSTALVC of the N-term of human alpha -smooth muscle actin (1) | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry, Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Dual RNAScope ISH-IHC | |
| 1A4 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| 59 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction