Learn More
alpha-Smooth Muscle Actin Antibody (SPM332), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34760AF405
Product Code. 18777533
  • € 584.00 / 0.10mL
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Baboon, Chimpanzee, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
ACTA2
Protein A or G purified
Store at 4C in the dark.
AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
42 kDa
Pig
IgG2a κ