missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (2A7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-05194-0.025ML
This item is not returnable.
View return policy
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Specifications
| alpha-Synuclein | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 | |
| P37840 | |
| SNCA | |
| Full length human recombinant protein with the epitope from amino acids 61-95 | |
| Immunogen affinity purified | |
| RUO | |
| 6622 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry (Free Floating), Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 2A7 | |
| Unconjugated | |
| 50% PBS, 50% Glycerol with 5mM Sodium Azide | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Mouse | |
| 15 kDa | |
| 0.025 mL | |
| Primary | |
| Pig, Human, Bovine, Rat, Mouse | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction