missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SCYL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29010
This item is not returnable.
View return policy
Description
SCYL2 Polyclonal antibody specifically detects SCYL2 in Human,Mouse samples. It is validated for Western Blot
Specifications
| SCYL2 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CVAK104, FLJ10074, KIAA1360Coated vesicle-associated kinase of 104 kDa, SCY1-like 2 (S. cerevisiae), SCY1-like protein 2 | |
| Between 825 and 875 | |
| 100 μg | |
| Protein Kinase | |
| 55681 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction