missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein, p Tyr136 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 479.00
Specifications
| Antigen | alpha-Synuclein |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638448
|
Novus Biologicals
NBP3-05404-100ul |
100 μg |
€ 479.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612429
|
Novus Biologicals
NBP3-05404-20ul |
20 μg |
€ 188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Synuclein Polyclonal antibody specifically detects alpha-Synuclein in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| alpha-Synuclein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 6622 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein | |
| A synthetic phosphorylated peptide around Y136 of human alpha-Synuclein (NP_000336.1). QDYEP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title