missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-TTP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92170-0.1ml
This item is not returnable.
View return policy
Description
Alpha-TTP Polyclonal antibody specifically detects Alpha-TTP in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Alpha-TTP | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| alpha-tocopherol transfer protein, alphaTTP, Alpha-TTP, ataxia (Friedreich-like) with vitamin E deficiency, ATTP, AVED, tocopherol (alpha) transfer protein, tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin Edeficiency), TPP1, TTP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human TTPA (NP_000361.1). MAEARSQPSAGPQLNALPDHSPLLQPGLAALRRRAREAGVPLAPLPLTDSFLLRFLRARDFDLDLAWRLLKNYYKWRAECPEISADLHPRSIIGLLKAGYHGVLRSRDPT | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 7274 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction