missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin 4a Antibody (B-5-1-2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-11304C
This item is not returnable.
View return policy
Description
alpha Tubulin 4a Monoclonal antibody specifically detects alpha Tubulin 4a in Human, Mouse, Rat, Bovine, Chicken, Primate samples. It is validated for Western Blot, Immunofluorescence
Specifications
| alpha Tubulin 4a | |
| Monoclonal | |
| DyLight 650 | |
| alpha 1 (testis specific), Alpha-tubulin 1, FLJ30169, H2-ALPHA, Testis-specific alpha-tubulin, TUBA1, Tubulin alpha-1 chain, tubulin alpha-4A chain, Tubulin H2-alpha, tubulin, alpha 1, tubulin, alpha 4a | |
| Sarkosyl-resistant filaments from Stronglycentrotus purpuratus (sea urchin). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 7277 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| B-5-1-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Chicken, Primate | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur