Learn More
alpha Tubulin Antibody (961258), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals MAB93441V
Product Code. 18597667
  • ONLINE EXCLUSIVE PRICE
    € 581.00
    € 549.15 / 100µL
    Save € 31.85
    5% Off
Description
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha Tubulin
Monoclonal
DyLight 405
Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific
This alpha Tubulin Antibody (961258) was developed against alpha Tubulin.
100 μg
Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience
7846
Store at 4°C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
961258
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat