missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (961258), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB93441IR
This item is not returnable.
View return policy
Description
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| alpha Tubulin | |
| Monoclonal | |
| DyLight 755 | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| This alpha Tubulin Antibody (961258) was developed against alpha Tubulin. | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| 7846 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 961258 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction