missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (SR1897), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | alpha Tubulin |
|---|---|
| Clone | SR1897 |
| Dilution | Western Blot 1:3000-1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691945
|
Novus Biologicals
NBP3-22671-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647094
|
Novus Biologicals
NBP3-22671-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Yeast samples. It is validated for Western BlotSpecifications
| alpha Tubulin | |
| Western Blot 1:3000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Yeast | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tublin, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| A synthesized peptide derived from yeast alpha Tubulin (Uniprot #: P09733) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1897 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7846 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title