missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUBA1A Polyclonal Antibody
GREENER_CHOICE

Product Code. 13423437
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13423437 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13423437 Supplier Invitrogen™ Supplier No. PA529444

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Jurkat, Raji, HeLa cytosol fraction, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat-2, Drosophila. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBA1A
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TUBA1A
Gene Accession No. P02552, P68369, P68370, Q71U36
Gene Alias alpha 1a antibody; alpha 3 antibody; alpha-tubulin; alpha-tubulin 1; Alpha-tubulin 3; alpha-tubulin 3 antibody; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; B-ALPHA-1 antibody; brain-specific antibody; Detyrosinated tubulin alpha-1A chain; FLJ25113 antibody; FLJ30169; H2 ALPHA; H2ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub1 antibody; hum-a-tub2; hum-a-tub2 antibody; LIS3; LIS3 antibody; LOC106557476; M40; MGC117247; MGC16435; OK/SW cl.56; OK/SWcl.56; OK/SW-cl.56; Talpha1 alpha-tubulin; TUBA1; Tuba-1; Tuba1a; TUBA1A antibody; TUBA3; TUBA3 antibody; TUBA6; TUBB1; TUBB5; tubulin alpha 1a; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-1A chain antibody; tubulin alpha-3 chain; tubulin alpha-3 chain antibody; Tubulin B-alpha-1; tubulin B-alpha-1 antibody; tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1A; tubulin, alpha 6; tubulin, alpha, brain-specific; unnamed protein product
Gene Symbols TUBA1A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human alpha Tubulin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22142, 429035, 64158, 7846
Target Species Chicken, Drosophila, Human, Insect, Mouse, Monkey, Protozoa, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
compliance-icons
Product Identifier
  • TUBA1A Polyclonal Antibody
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Glycerin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.