missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody (1H3.B8), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12875UV
This item is not returnable.
View return policy
Description
AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| AlphaA Crystallin/CRYAA | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CRYAA | |
| Native Alpha Crystallin | |
| 200 μL | |
| Primary | |
| Detects approx 20kDa. Does not cross-react with aB-crystallin, Beta L-crystallin, BH- crystallin, gamma-crystallin, HSP25, HSP27 or HSP47 proteins. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1H3.B8 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
| Mouse | |
| Protein G purified | |
| Membrane Trafficking and Chaperones, Vision | |
| 1409 | |
| Human, Mouse, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction