missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody (SR1256), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | AlphaA Crystallin/CRYAA |
|---|---|
| Clone | SR1256 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654355
|
Novus Biologicals
NBP3-22159-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696814
|
Novus Biologicals
NBP3-22159-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AlphaA Crystallin/CRYAA Monoclonal antibody specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| AlphaA Crystallin/CRYAA | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
| A synthesized peptide derived from human AlphaA Crystallin/CRYAA (Uniprot #: P02489) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1256 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Vision | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1409 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title