missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (3A10.C9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22410AF488
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat, Bovine samples. It is validated for ELISA, Immunohistochemistry, Microarray
Specifications
| AlphaB Crystallin/CRYAB | |
| Monoclonal | |
| Alexa Fluor 488 | |
| P02511, NP_001876.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine | |
| Purified |
| ELISA, Immunohistochemistry, Microarray | |
| 3A10.C9 | |
| 50mM Sodium Borate | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Native Alpha B Crystallin | |
| 0.2 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
| 1410 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction