missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2519AF532
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Polyclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| AlphaB Crystallin/CRYAB | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
| 1410.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CRYAB | |
| A synthetic peptide made to an internal region of the human Crystallin AB protein (within residues 70-140). [Swiss-Prot P02511] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction