missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (OTI6A9), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70454APCCY7
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Monoclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| AlphaB Crystallin/CRYAB | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CRYAB | |
| Full length recombinant protein of human AlphaB Crystallin/CRYAB (NP_001876) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI6A9 | |
| Flow Cytometry | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
| 1410.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction