Learn More
AlphaB Crystallin/CRYAB Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-2519PCP
Product Code. 18568660
  • € 585.00 / 0.10mL
Description
AlphaB Crystallin/CRYAB Polyclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence