Description
- Specialized protection: The AlphaTec™ 1500 PLUS Stitched - Model 111 delivers light chemical resistance, guards against asbestos and filters 99.9% of particles over 3 microns in size, as well as brick and cement dust, light aerosols and mists
- Heightened breathability: This asbestos protection suit's air and water vapor permeability also reduces wearers' exposure to heat stress
- Enhanced fit and features: Wearer comfort is augmented by this protective body suit's optimal fit, while a three-piece hood offers additional protective coverage, finger loops offer a secure hold and red stitching ensures easy identification
- Certified antistatic capabilities: The AlphaTec™ 1500 PLUS Stitched - Model 111 is tested against EN 1149-5 antistatic standards
Specifications
Specifications
| Size | Small |
| Gender | Unisex |
| Material | Anti-static SMS |
| Color | Blue |
| Disposable | Reusable |
| Closure Type | 2-way Zipper with Storm Flap |
| Sterility | Non-sterile |
| Protection Class | CE CATEGORY III (Type 5, Type 6) |
| Resistance | Particle, Anti-static, Chemicals |
| Collar Type | Elasticated hood |
| Show More |