missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2300 STANDARD Bound
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Y23B103
Y23B111
Unit Size:
Pack of 40
Description
- Lightweight, yet relatively strong and durable
- Designed to protect, typical coverall features include respirator fit hood and a zip flap with self-adhesive tape closure
- Anti-static
Specifications
Specifications
| Size | Medium |
| Material | Polyethylene on Bicomponent Polypropylene, Polyethylene Non-Woven |
| Color | Yellow |
| Seam Type | Bound |
| Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
| For Use With (Application) | Environmental Clean-up and Remediation, Assembly and Disassembly of Large Pipes, Transferring Liquids and Solids, Assembling and Mounting Components |
| Weight (Metric) | 65 gsm |
| Style | Y23B103 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction