missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 Overall with Hood, Yellow
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 15
Pack of 20
Description
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Lightweight and durable
- Highly visible bright yellow color for improved worker safety
- Anti-static
Specifications
Specifications
| Size | 4X-Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Seam Type | Welded, Taped |
| Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
| Model | 162 |
| For Use With (Application) | Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Unexpected Leakages Spills or Other Releases, Opening and Draining Pumps Valves or Lines, Loading and Unloading Trucks and Vehicles, Filling, Blending and Charging of Raw Materials, Product Inspection, Mixing or Loading Chemicals, Cleaning of Plant and Machines |
| Weight (Metric) | 80 gsm |
| Style | Y30T162 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction