missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 Jacket
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect
- Silicone free
Specifications
Specifications
| Size | Small |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Green |
| Closure Type | Dual Zipper |
| Seam Type | Welded and Taped |
| Certifications/Compliance (Europe) | Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003 |
| Product Type | Chemical-resistant Jacket |
| Style | G40T230 |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?