missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 5000
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
O50T103
O50T111
O50T122
O50T125B
O50T151
O50T151B
Unit Size:
Pack of 10
Pack of 8
Description
- Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
- Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Designed to protect, rear entry suit with neoprene rubber face seal for a close fit to full face respirators
- Welded and taped seams, our highest barrier to liquids and particulates
- Designed to protect, typical innovative features include dual zip systems, double cuffs with knitted inner cuffs
- Multi-layer barrier material which is lightweight, yet strong and durable
- Anti-static
Specifications
Specifications
| Size | 2X-Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Orange |
| Seam Type | Welded, Taped |
| Certifications/Compliance (Europe) | EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases |
| Weight (Metric) | 132 gsm |
| Style | O50T125B |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction