missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALPL Monoclonal Antibody (W8B2B10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546185
This item is not returnable.
View return policy
Description
This antibody recognizes TNAP (tissue non-specific alkaline phosphatase), an ectoenzyme expressed mainly on embryonic stem cells, liver, bone, and kidney cells. This antibody is suitable for characterization of bone marrow-derived MSCs, iPSCs, and ESCs.
Alkaline phosphatase is a homodimeric protein enzyme (86 kDa), containing two zinc atoms crucial to its catalytic function per monomer, and it is optimally active at alkaline pH environments. The enzyme is found in prokaryotes and eukaryotes, with the same general function but in different structural forms suitable to organism's environment. ALP plays a role in liver metabolism and development within the skeleton. In bacteria, ALP acts as a source of inorganic phosphate. In yeast, ALP can be a useful protein for studying membrane protein sorting.
Specifications
| ALPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P05186 | |
| ALPL | |
| WERI-RB-1 retinoblastoma cell line. | |
| 100 μg | |
| Primary | |
| Human, Canine, Monkey, Sheep, Pig, Goat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| W8B2B10 | |
| Unconjugated | |
| ALPL | |
| Akp2; Akp-2; Alkaline phosphatase 2; alkaline phosphatase 2, liver; alkaline phosphatase liver/bone/kidney isozyme; alkaline phosphatase precursor (EC 3.1.3.2); alkaline phosphatase tissue non-specific isoform; alkaline phosphatase tissue non-specific isoform/TNS-AP; alkaline phosphatase, biomineralization associated; alkaline phosphatase, intestinal; alkaline phosphatase, liver/bone/kidney; alkaline phosphatase, tissue-nonspecific isozyme; alkaline phosphomonoesterase; ALPI; Alpl; APTNAP; AP-TNAP; FLJ40094; FLJ93059; glycerophosphatase; HOPS; IAP; intestinal alkaline phosphatase; intestinal-type alkaline phosphatase; Kasahara isozyme; liver/bone/kidney isozyme; liver/bone/kidney-type alkaline phosphatase; MGC161443; MGC167935; PHOA; tissue-nonspecific alkaline phosphatase; tissue-nonspecific alkaline phosphatase precursor; tissue-nonspecific ALP; Tissue-nonspecific ALP alkaline phosphatase; TNAP; TNSALP; TNS-AP | |
| Mouse | |
| Protein A | |
| RUO | |
| 100170147, 101123195, 102189447, 249, 403548 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction