missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527667
This item is not returnable.
View return policy
Description
Recommended positive controls: THP-1. Predicted reactivity: Pig (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a serum protein that binds insulin-like growth factors, increasing their half-life and their vascular localization. Production of the encoded protein, which contains twenty leucine-rich repeats, is stimulated by growth hormone. Defects in this gene are a cause of acid-labile subunit deficiency, which maifests itself in a delayed and slow puberty. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| ALS | |
| Polyclonal | |
| Unconjugated | |
| IGFALS | |
| ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P35858 | |
| IGFALS | |
| Recombinant protein encompassing a sequence within the C-terminus region of human ALS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction