missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALS2CR11 Monoclonal Antibody (OTI7A8)
GREENER_CHOICE

Product Code. 15977084
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15977084 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15977084 Supplier Invitrogen™ Supplier No. MA527497

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALS2CR11 Monoclonal Antibody for Western Blot

ALS2CR11 has 2 isoforms of the human protein that are produced by alternative splicing. An autosomal recessive form of juvenile amyotrophic lateral sclerosis was originally mapped to a region of chromosome 2 that includes this gene. The encoded protein contains a calcium-dependent membrane targeting C2 domain. This domain is often found in proteins that are involved in membrane trafficking and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS2CR11
Applications Western Blot
Classification Monoclonal
Clone OTI7A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C2CD6
Gene Accession No. Q53TS8
Gene Alias 1700052H20Rik; 4930408G06Rik; Als2cr11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 11 (human); amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein; amyotrophic lateral sclerosis 2 chromosome region candidate 11; C2 calcium-dependent domain-containing protein 6; C2CD6; testicular tissue protein Li 16
Gene Symbols C2CD6
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 307-623 of ALS2CR11 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151254
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.