missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALS2CR8 Antibody (2A3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079800-M01
This item is not returnable.
View return policy
Description
ALS2CR8 Monoclonal antibody specifically detects ALS2CR8 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA, KnockDown
Specifications
| ALS2CR8 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 8, amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein, Calcium-response factor, CaRF, CARFDKFZp667N246, FLJ12675, FLJ21579, NYD-SP24, Testis development protein NYD-SP24 | |
| ALS2CR8 (NP_079020, 621 a.a. ∽ 718 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CLTQNNSTASTMGNLPEPDQNLVAMDELVEVGDVEDTGNLEGTVHRILLGDVQTIPIQIIDNHSALIEENPESTISVSQVKQEPKEPALSMEAKNCGL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2A3 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA, Knockdown Validated | |
| NP_079020 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 79800 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction