missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alsin Polyclonal Antibody
GREENER_CHOICE

Product Code. 16660045 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16660045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16660045 Supplier Invitrogen™ Supplier No. PA5103595

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ALS2.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alsin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ALS2
Gene Accession No. P0C5Y8, Q920R0, Q96Q42
Gene Alias 3222402C23Rik; 9430073A21Rik; Als2; ALS2, alsin Rho guanine nucleotide exchange factor; Als2cr6; Alsin; ALSJ; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; IAHSP; KIAA1563; MGC87187; mKIAA1563; PLSJ
Gene Symbols ALS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALS2(Accession Q96Q42), corresponding to amino acid residues L204-S254.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363235, 57679, 74018
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.