Learn More
Macherey-Nagel™ Alugram Aluminum Sheet CEL 300 UV254
€ 102.00 - € 160.00
Specifications
Type | Aluminum Sheet |
---|---|
Fluorescent Indicator | UV254 |
For Use With (Application) | Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates |
Specific Surface | 15,000 cm2/g |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10338651
|
Macherey-Nagel™
818156 |
20 x 20 cm |
€ 160.00
Pack of 25 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
10516881
|
Macherey-Nagel™
818157 |
5 x 2 cm |
€ 102.00
Pack of 50 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Specifications
Aluminum Sheet | |
UV254 | |
Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates | |
15,000 cm2/g |