Learn More
Macherey-Nagel™ Alugram Aluminum Sheet Nano-SIL G
Product Code. 12770753 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Product Code. 12770753
Brand: Macherey-Nagel™ 818240
Only null left
Specifications
Particle Size 2 to 10 μm
Pore Size 60 Å
Quantity 50 Pk.
Type Aluminum Sheet
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 5 x 20 cm