Learn More
Lovibond™ Aluminum #2 tablets
ECR method for comparators and photometers
€ 42.00 - € 86.00
Specifications
Range Aluminum #2 0.01 to 0.30 mg/L
Type Reagent
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13161076
View Documents Promotion Details
Lovibond™
515470BT
100 Tablets
€ 42.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
13171076
View Documents Promotion Details
Lovibond™
515471BT
250 Tablets
€ 86.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Description
Description
ECR method for comparators and photometers
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
Specifications
Aluminum #2 0.01 to 0.30 mg/L
Reagent