missing translation for 'onlineSavingsMsg'
Learn More

Aluminum AA Standard: Al @ 1000 μg/mL in 5% HCl, VHG™

Product Code. 17902636
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 17902636

Brand: VHG Labs VHGAAALH500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Aluminum AA Standard: Al @ 1000 μg/mL in 5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium
CAS 7429-90-5
Methods AA/GFAA
Analyte or Component Names Aluminium
Shipping Condition Room Temperature
Molecular Formula Al
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Aluminium
InChI Formula InChI=1S/Al
SMILES [Al]
IUPAC Name Aluminum
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 26.98
Quantity 500 mL
Formula Weight 26.9815 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.