Learn More
Bochem™ Aluminum Bossheads
Product Code. 10751392
Click to view available options
Diameter (Metric) Outer:
16.2 mm
16.5 mm
27 mm
Thread Size:
8
10
Weight (Metric):
0.06 kg
0.09 kg
0.13 kg
0.3 kg
Unit Size:
Each
Product Code. 10751392
Brand: Bochem™ 5350
Only null left
Add to basket
Add to basket
Manufactured from aluminum. Bochem™ Aluminum Bossheads are DIN 12895 certified.
  • Manufactured from aluminum
  • DIN 12895
  • Type 1: Thumb screw M8
Specifications
Material Aluminum
Diameter (Metric) Outer 16.5 mm
Product Type Aluminum Bossheads
Thread Size 8
Quantity 1 Ea.
Weight (Metric) 0.13 kg