Learn More
Aluminum Bromide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Aluminum Bromide |
| CAS | 7727-15-3 |
| Molecular Formula | AlBr3 |
| Quantity | 25 g |
| Synonym | Aluminum Tribromide, Aluminum Tribromide (AlBr3), NSC 87608 |
| InChI Formula | InChI=1S/Al.3BrH/h;3*1H/q+3;;;/p-3 |
| SMILES | Br[Al](Br)Br |
| IUPAC Name | tribromoalumane |
| Molecular Weight (g/mol) | 266.69 |
| Formula Weight | 263.7366 |