Learn More
Haldenwanger™ Aluminum Ceramic Conical Tall Form Crucibles
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
45 mL
250 mL
Unit Size:
Each
Specifications
Specifications
| Material | Aluminum Ceramic |
| Capacity (Metric) | 20 mL |
| Product Type | Crucible |
| Shape | Conical |