Learn More
Bochem™ Aluminum Chemical Scoop
Click to view available options
Capacity (Metric):
10 mL
65 mL
90 mL
240 mL
400 mL
480 mL
900 mL
1.6 L
Length (Metric):
52 mm
145 mm
155 mm
250 mm
270 mm
285 mm
290 mm
310 mm
350 mm
Unit Size:
Each
Description
- Lightweight
- Aluminum
- Length is total for combined scoop and handle
Specifications
Specifications
| Capacity (Metric) | 900 mL |
| Product Type | Chemical Scoop |
| Material | Aluminum |
| Length (Metric) | 350 mm |