Learn More
New Brunswick Scientific™ Aluminum Erlenmeyer Flask Platform
Product Code. 11622744
Click to view available options
For Use With (Equipment):
Innova 44/44R Incubator Shaker
Unit Size:
Each
Product Code. 11622744
Brand: New Brunswick Scientific™ M12829907
Only null left
Add to basket
Add to basket
Use these trays with Innova™ 44/44R stackable incubator shakers. New Brunswick Scientific™ Aluminum Erlenmeyer Flask Platforms are designed for 500mL Erlenmeyer flasks
  • Aluminum Erlenmeyer flask dedicated platform
  • 76 × 46cm
  • Not interchangeable with other 76 × 46cm platforms
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Dimensions (L x W) 76 x 46 cm
For Use With (Equipment) Innova 44/44R Incubator Shaker