Learn More
Brady™ Aluminum Mandatory Signs
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
Displayed Text:
Cotton Blouse
Hearing Protection
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | NF X 08-003-1 and 08-003-3 NX and the European Directive 92/58 / EEC |
| Alternate Part Number | PIC 2209-DIA 200-B7525 FF |
| Material | Aluminum |
| Quantity | 1 Ea. |
| Diameter (Metric) | 200 mm |
| For Use With (Application) | Indoor and outdoor safety signs |
| Shape | Round |
| Displayed Text | Cotton Blouse |
| Product Type | Mandatory Sign |
| Color | White on Blue |