Learn More
Sampling Systems™ Aluminum Multilevel Slot Sampler
Tous les produits Sampling Systems Produits
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
880 mL
1.7 L
2.9 L
Length (Metric):
900 mm
1500 mm
2500 mm
Includes:
3 Slots
5 Slots
7 Slots
missing translation for 'unitSize'
Each
Description
- Made from aluminum
- Cross section (Composite sample)
- Suitable for taking large volume samples
- aluminum samplers are not suitable for use with GMP products
Spécification
Spécification
| Material | Aluminum |
| Description | Slot Sampler |
| Includes | 5 Slots |
| Capacity (Metric) | 1.7 L |
| Diameter (Metric) Outer | 50 mm |
| Length (Metric) | 1500 mm |